siRNA Details
| Gene Symbol: Ppapdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293703.1 | NP_001280632.1 | 2, 3 | 209 | ||
| NM_028000.1 | NP_082276.1 | 2, 3 | 209 | |||
| GenBank mRNA | AA184457.1 | - | 2, 3 | 209 | ||
| AK009455.1 | - | 2, 3 | 176 | |||
| AK144593.1 | - | 2, 3 | 194 | |||
| BB857717.1 | - | 2, 3 | 210 | |||
| BC099489.1 | - | 2, 3 | 160 | |||

Ppapdc1b
Genomic Map