siRNA Details
| Gene Symbol: Ppapdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293703.1 | NP_001280632.1 | 8 | 1458 | ||
| NM_028000.1 | NP_082276.1 | 7 | 1375 | |||
| GenBank mRNA | AK007542.1 | - | 4 | 1979 | ||
| AK144593.1 | - | 7 | 1360 | |||
| BC023111.1 | - | 7 | 1320 | |||
| BC039626.1 | - | 6 | 1190 | |||
| BC062173.1 | - | 7 | 1308 | |||
| BC089556.1 | - | 7 | 1277 | |||
| BC099489.1 | - | 8 | 1409 | |||

Ppapdc1b
Genomic Map