siRNA Details
| Gene Symbol: Zcchc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101433.1 | NP_001094903.1 | 4 | 2233 | ||
| GenBank mRNA | BC034735.1 | - | 2 | 1787 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcchc24| Gene Symbol: Zcchc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101433.1 | NP_001094903.1 | 4 | 2233 | ||
| GenBank mRNA | BC034735.1 | - | 2 | 1787 | ||