siRNA Details
| Gene Symbol: Tom1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028011.2 | NP_082287.1 | 15 | 1450 | ||
| GenBank mRNA | AF395837.1 | - | 15 | 1414 | ||
| AJ505007.1 | - | 15 | 1411 | |||
| AK009826.1 | - | 13 | 1185 | |||
| AK033712.1 | - | 15 | 1452 | |||
| AK083630.1 | - | 1412 | ||||
| BC004710.1 | - | 8 | 1807 | 3499918 | ||

Tom1l1
Genomic Map