siRNA Details
| Gene Symbol: 2410012M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028033.1 | NP_082309.1 | 1 | 984 | ||
| GenBank mRNA | AK010476.1 | - | 1 | 963 | ||
| BC128017.1 | - | 1 | 953 | 40136125 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2410012M07Rik| Gene Symbol: 2410012M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028033.1 | NP_082309.1 | 1 | 984 | ||
| GenBank mRNA | AK010476.1 | - | 1 | 963 | ||
| BC128017.1 | - | 1 | 953 | 40136125 | ||