siRNA Details
| Gene Symbol: Lmntd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028050.2 | NP_082326.1 | 11 | 1475 | ||
| GenBank mRNA | AK005476.1 | - | 2 | 62 | ||
| BC119093.1 | - | 11 | 1479 | 8733842 | ||
| BC119119.1 | - | 11 | 1479 | 8733868 | ||
| BC144834.1 | - | 11 | 1509 | 9053382 | ||

Lmntd2
Genomic Map