siRNA Details
| Gene Symbol: Cyb561d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081320.2 | NP_001074789.2 | 3 | 3235 | ||
| NM_028061.1 | NP_082337.1 | 3 | 3102 | |||
| GenBank mRNA | AK087683.1 | - | 3 | 3189 | ||
| BC043107.1 | - | 3 | 3721 | |||
| BC060107.1 | - | 3 | 3077 | |||

Cyb561d1
Genomic Map