siRNA Details
| Gene Symbol: 2010001E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163503.1 | NP_001156975.1 | 4 | 1488 | ||
| GenBank mRNA | AK008003.1 | - | 4 | 1489 | ||
| BC145051.1 | - | 1656 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2010001E11Rik| Gene Symbol: 2010001E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163503.1 | NP_001156975.1 | 4 | 1488 | ||
| GenBank mRNA | AK008003.1 | - | 4 | 1489 | ||
| BC145051.1 | - | 1656 | ||||