siRNA Details
| Gene Symbol: 2010111I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289924.1 | NP_001276853.1 | 8 | 2021 | ||
| NM_001289926.1 | NP_001276855.1 | 8 | 2021 | |||
| GenBank mRNA | AJ810420.1 | - | 7 | 1750 | ||
| AK008397.1 | - | 4 | 480 | |||
| AK141931.1 | - | 7 | 1987 | |||
| BC042715.1 | - | 7 | 1439 | |||
| BC141279.1 | - | 7 | 1872 | 9055891 | ||

2010111I01Rik
Genomic Map