siRNA Details
| Gene Symbol: 2010300C02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028096.1 | NP_082372.1 | 10 | 3871 | ||
| GenBank mRNA | AK008485.1 | - | 4 | 1795 | ||
| BC029089.1 | - | 4 | 2049 | |||
| BC072639.1 | - | 10 | 3871 | 30355303 | ||

2010300C02Rik
Genomic Map