siRNA Details
| Gene Symbol: Klhl35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028145.1 | NP_082421.1 | 4 | 1204 | ||
| GenBank mRNA | AK013012.1 | - | 4 | 1182 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl35| Gene Symbol: Klhl35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028145.1 | NP_082421.1 | 4 | 1204 | ||
| GenBank mRNA | AK013012.1 | - | 4 | 1182 | ||