siRNA Details
Gene Symbol: Klhl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028145.1 | NP_082421.1 | 7 | 1937 | ||
GenBank mRNA | AK013012.1 | - | 7 | 1915 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klhl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028145.1 | NP_082421.1 | 7 | 1937 | ||
GenBank mRNA | AK013012.1 | - | 7 | 1915 |