siRNA Details
Gene Symbol: Zfp558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028935.1 | NP_083211.1 | 8 | 1617 | ||
GenBank mRNA | AK005691.1 | - | 2 | 842 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028935.1 | NP_083211.1 | 8 | 1617 | ||
GenBank mRNA | AK005691.1 | - | 2 | 842 |