siRNA Details
| Gene Symbol: 1600014C23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028164.1 | NP_082440.1 | 1 | 93 | ||
| GenBank mRNA | AK005450.1 | - | 1 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1600014C23Rik| Gene Symbol: 1600014C23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028164.1 | NP_082440.1 | 1 | 93 | ||
| GenBank mRNA | AK005450.1 | - | 1 | 94 | ||