siRNA Details
| Gene Symbol: 1810041L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163145.1 | NP_001156617.1 | 5 | 5254 | ||
| GenBank mRNA | AK162836.1 | - | 1 | 828 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810041L15Rik| Gene Symbol: 1810041L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163145.1 | NP_001156617.1 | 5 | 5254 | ||
| GenBank mRNA | AK162836.1 | - | 1 | 828 | ||