siRNA Details
Gene Symbol: Klhl40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028202.3 | NP_082478.1 | 1 | 53 | ||
GenBank mRNA | BC125576.1 | - | 1 | 53 | 40130091 | |
BC137889.1 | - | 1 | 53 | 8860908 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klhl40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028202.3 | NP_082478.1 | 1 | 53 | ||
GenBank mRNA | BC125576.1 | - | 1 | 53 | 40130091 | |
BC137889.1 | - | 1 | 53 | 8860908 |