siRNA Details
| Gene Symbol: Dnajc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033165.3 | NP_001028337.2 | 4 | 1447 | ||
| GenBank mRNA | BM951172.1 | - | 1 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc25| Gene Symbol: Dnajc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033165.3 | NP_001028337.2 | 4 | 1447 | ||
| GenBank mRNA | BM951172.1 | - | 1 | 93 | ||