siRNA Details
| Gene Symbol: Dnajc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033165.3 | NP_001028337.2 | 3 | 541 | ||
| GenBank mRNA | AK008359.1 | - | 4 | 295 | ||
| AK008425.1 | - | 4 | 571 | |||
| AK136359.1 | - | 3 | 235 | |||
| BC119039.1 | - | 3 | 306 | 8733788 | ||
| BC120693.1 | - | 3 | 306 | 40129616 | ||

Dnajc25
Genomic Map