siRNA Details
| Gene Symbol: Dnajc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033165.3 | NP_001028337.2 | 3 | 978 | ||
| GenBank mRNA | AK136359.1 | - | 3 | 672 | ||
| BC119039.1 | - | 3 | 743 | 8733788 | ||
| BC120693.1 | - | 3 | 743 | 40129616 | ||
| BX512063.1 | - | 1 | 268 | |||

Dnajc25
Genomic Map