siRNA Details
Gene Symbol: Resp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019278.1 | NP_062151.1 | 2, 3 | 306 | ||
GenBank mRNA | BC058147.1 | - | 2, 3 | 302 | 6922403 | |
L25633.1 | - | 2, 3 | 306 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Resp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019278.1 | NP_062151.1 | 2, 3 | 306 | ||
GenBank mRNA | BC058147.1 | - | 2, 3 | 302 | 6922403 | |
L25633.1 | - | 2, 3 | 306 |