siRNA Details
Gene Symbol: Spink5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081180.1 | NP_001074649.1 | 32 | 4128 | ||
GenBank mRNA | AK010042.1 | - | 5 | 1453 | ||
AK028727.1 | - | 13 | 2278 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spink5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081180.1 | NP_001074649.1 | 32 | 4128 | ||
GenBank mRNA | AK010042.1 | - | 5 | 1453 | ||
AK028727.1 | - | 13 | 2278 |