siRNA Details
Gene Symbol: Spink5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081180.1 | NP_001074649.1 | 29 | 2825 | ||
GenBank mRNA | AJ426449.2 | - | 29 | 2823 | ||
AK010042.1 | - | 2 | 151 | |||
AK028727.1 | - | 10 | 975 | |||
BC118011.1 | - | 29 | 2797 | 40097427 |
