siRNA Details
| Gene Symbol: Spink5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081180.1 | NP_001074649.1 | 10 | 937 | ||
| GenBank mRNA | AJ426449.2 | - | 10 | 935 | ||
| AK035690.1 | - | 10 | 938 | |||
| AK036804.1 | - | 10 | 938 | |||
| AK162690.1 | - | 10 | 938 | |||
| BC118011.1 | - | 10 | 909 | 40097427 | ||

Spink5
Genomic Map