siRNA Details
Gene Symbol: Cwf19l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081077.1 | NP_001074546.1 | 14 | 2664 | ||
GenBank mRNA | AK088543.1 | - | 16 | 3906 | ||
AK139654.1 | - | 14 | 2665 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cwf19l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081077.1 | NP_001074546.1 | 14 | 2664 | ||
GenBank mRNA | AK088543.1 | - | 16 | 3906 | ||
AK139654.1 | - | 14 | 2665 |