siRNA Details
| Gene Symbol: Taf4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100449.1 | NP_001093919.1 | 15 | 3490 | ||
| GenBank mRNA | AK029943.1 | - | 4 | 1035 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Taf4b| Gene Symbol: Taf4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100449.1 | NP_001093919.1 | 15 | 3490 | ||
| GenBank mRNA | AK029943.1 | - | 4 | 1035 | ||