siRNA Details
| Gene Symbol: Wdr43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175639.1 | NP_783570.1 | 18 | 2540 | ||
| GenBank mRNA | AK129031.1 | - | 18 | 2458 | ||
| AK152321.1 | - | 18 | 2530 | |||
| BC079913.1 | - | 17 | 2241 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr43| Gene Symbol: Wdr43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175639.1 | NP_783570.1 | 18 | 2540 | ||
| GenBank mRNA | AK129031.1 | - | 18 | 2458 | ||
| AK152321.1 | - | 18 | 2530 | |||
| BC079913.1 | - | 17 | 2241 | |||