siRNA Details
| Gene Symbol: Shisa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174086.1 | NP_001167557.1 | 4 | 2271 | ||
| NM_028277.2 | NP_082553.2 | 5 | 2319 | |||
| GenBank mRNA | AK049860.1 | - | 5 | 1845 | ||
| BC030884.1 | - | 4 | 1560 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shisa9| Gene Symbol: Shisa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174086.1 | NP_001167557.1 | 4 | 2271 | ||
| NM_028277.2 | NP_082553.2 | 5 | 2319 | |||
| GenBank mRNA | AK049860.1 | - | 5 | 1845 | ||
| BC030884.1 | - | 4 | 1560 | |||