siRNA Details
| Gene Symbol: Zfp518a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028319.1 | NP_082595.1 | 5 | 5210 | ||
| GenBank mRNA | AK220334.1 | - | 1 | 1526 | ||
| BC138382.1 | - | 3 | 4916 | 8861402 | ||
| BC138383.1 | - | 3 | 4916 | 8861403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp518a| Gene Symbol: Zfp518a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028319.1 | NP_082595.1 | 5 | 5210 | ||
| GenBank mRNA | AK220334.1 | - | 1 | 1526 | ||
| BC138382.1 | - | 3 | 4916 | 8861402 | ||
| BC138383.1 | - | 3 | 4916 | 8861403 | ||