siRNA Details
| Gene Symbol: Grrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099296.2 | NP_001092766.1 | 1 | 47 | ||
| GenBank mRNA | BC048704.1 | - | 1 | 43 | 6330338 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grrp1| Gene Symbol: Grrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099296.2 | NP_001092766.1 | 1 | 47 | ||
| GenBank mRNA | BC048704.1 | - | 1 | 43 | 6330338 | |