siRNA Details
| Gene Symbol: Zfp618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028326.1 | NP_082602.1 | 15 | 2442 | ||
| GenBank mRNA | AK012894.1 | - | 1 | 181 | ||
| AK173309.1 | - | 1 | 689 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp618| Gene Symbol: Zfp618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028326.1 | NP_082602.1 | 15 | 2442 | ||
| GenBank mRNA | AK012894.1 | - | 1 | 181 | ||
| AK173309.1 | - | 1 | 689 | |||