siRNA Details
| Gene Symbol: 2810459M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144992.1 | NP_001138464.1 | 3 | 900 | ||
| NM_001144993.1 | NP_001138465.1 | 3 | 897 | |||
| GenBank mRNA | AK013367.1 | - | 3 | 496 | ||
| BC137675.1 | - | 3 | 897 | 8860694 | ||
| BC144821.1 | - | 3 | 900 | 9053369 | ||

2810459M11Rik
Genomic Map