siRNA Details
| Gene Symbol: 2810459M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144992.1 | NP_001138464.1 | 3 | 2745 | ||
| NM_001144993.1 | NP_001138465.1 | 3 | 2742 | |||
| GenBank mRNA | AK013367.1 | - | 3 | 2343 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810459M11Rik| Gene Symbol: 2810459M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144992.1 | NP_001138464.1 | 3 | 2745 | ||
| NM_001144993.1 | NP_001138465.1 | 3 | 2742 | |||
| GenBank mRNA | AK013367.1 | - | 3 | 2343 | ||