siRNA Details
| Gene Symbol: Zfp839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199785.1 | NP_001186714.1 | 7 | 2527 | ||
| NM_028365.2 | NP_082641.1 | 8 | 2755 | |||
| GenBank mRNA | BC150732.1 | - | 8 | 2755 | 9087643 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp839| Gene Symbol: Zfp839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199785.1 | NP_001186714.1 | 7 | 2527 | ||
| NM_028365.2 | NP_082641.1 | 8 | 2755 | |||
| GenBank mRNA | BC150732.1 | - | 8 | 2755 | 9087643 | |