siRNA Details
| Gene Symbol: Hspb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028394.2 | NP_082670.1 | 3 | 267 | ||
| GenBank mRNA | AK013636.1 | - | 3 | 244 | ||
| AK157651.1 | - | 3 | 268 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspb11| Gene Symbol: Hspb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028394.2 | NP_082670.1 | 3 | 267 | ||
| GenBank mRNA | AK013636.1 | - | 3 | 244 | ||
| AK157651.1 | - | 3 | 268 | |||