siRNA Details
| Gene Symbol: Abracl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028440.1 | NP_082716.1 | 3 | 610 | ||
| GenBank mRNA | AK013984.1 | - | 3 | 572 | ||
| AK171138.1 | - | 3 | 513 | |||
| BC039801.1 | - | 3 | 489 | |||
| BC098509.1 | - | 3 | 552 | |||
| DV053786.1 | - | 3 | 659 | |||

Abracl
Genomic Map