siRNA Details
| Gene Symbol: 3110043O21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081343.1 | NP_001074812.1 | 2, 3 | 588 | ||
| GenBank mRNA | AK154817.1 | - | 2, 3 | 562 | ||
| AK167354.1 | - | 2, 3 | 519 | |||
| BC076612.1 | - | 2, 3 | 690 | 30638238 | ||

3110043O21Rik
Genomic Map