siRNA Details
| Gene Symbol: 3110043O21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081343.1 | NP_001074812.1 | 11 | 3107 | ||
| GenBank mRNA | AK150973.1 | - | 1 | 1366 | ||
| AK167354.1 | - | 11 | 3037 | |||
| BC026738.1 | - | 9 | 2512 | 4020832 | ||
| BC076612.1 | - | 10 | 3325 | 30638238 | ||

3110043O21Rik
Genomic Map