siRNA Details
Gene Symbol: 3110052M02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166497.1 | NP_001159969.1 | 6 | 2939 | ||
GenBank mRNA | BC064464.1 | - | 3 | 2759 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 3110052M02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166497.1 | NP_001159969.1 | 6 | 2939 | ||
GenBank mRNA | BC064464.1 | - | 3 | 2759 |