siRNA Details
| Gene Symbol: Mrgbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028479.1 | NP_082755.1 | 6 | 909 | ||
| GenBank mRNA | AK005551.1 | - | 5 | 549 | ||
| AK038431.1 | - | 4 | 536 | |||
| AK217601.1 | - | 4 | 326 | |||
| BC042508.1 | - | 3 | 1715 | |||
| BC054431.1 | - | 4 | 805 | |||
| BC085320.1 | - | 2 | 2208 | |||
| BC110993.1 | - | 5 | 515 | 5363496 | ||
| BC116732.1 | - | 6 | 909 | 40126051 | ||
| BC116734.1 | - | 6 | 909 | 40126053 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001173739, XM_008762593, CO403993, BC099155.

Mrgbp
Genomic Map