siRNA Details
| Gene Symbol: Cib4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028483.1 | NP_082759.1 | 1 | 48 | ||
| GenBank mRNA | AK006670.1 | - | 1 | 49 | ||
| BC118938.1 | - | 1 | 13 | 40091772 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cib4| Gene Symbol: Cib4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028483.1 | NP_082759.1 | 1 | 48 | ||
| GenBank mRNA | AK006670.1 | - | 1 | 49 | ||
| BC118938.1 | - | 1 | 13 | 40091772 | ||