siRNA Details
| Gene Symbol: 1700031F05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028496.1 | NP_082772.1 | 7 | 1151 | ||
| GenBank mRNA | AK006578.1 | - | 7 | 1152 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700031F05Rik| Gene Symbol: 1700031F05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028496.1 | NP_082772.1 | 7 | 1151 | ||
| GenBank mRNA | AK006578.1 | - | 7 | 1152 | ||