siRNA Details
| Gene Symbol: 1700031F05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028496.1 | NP_082772.1 | 2 | 188 | ||
| GenBank mRNA | AK006578.1 | - | 2 | 189 | ||
| Gene Symbol: Gm5166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC049655.1 | - | 2 | 128 | ||

1700031F05Rik
Genomic Map