siRNA Details
| Gene Symbol: Itpripl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163527.1 | NP_001156999.1 | 2 | 2257 | ||
| NM_001163528.1 | NP_001157000.1 | 2 | 2081 | |||
| GenBank mRNA | AK028913.1 | - | 3 | 1997 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itpripl1| Gene Symbol: Itpripl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163527.1 | NP_001156999.1 | 2 | 2257 | ||
| NM_001163528.1 | NP_001157000.1 | 2 | 2081 | |||
| GenBank mRNA | AK028913.1 | - | 3 | 1997 | ||