siRNA Details
| Gene Symbol: Cldn34b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081671.1 | NP_001075140.1 | 2 | 149 | ||
| GenBank mRNA | BC100353.1 | - | 2 | 149 | 30933439 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cldn34b2| Gene Symbol: Cldn34b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081671.1 | NP_001075140.1 | 2 | 149 | ||
| GenBank mRNA | BC100353.1 | - | 2 | 149 | 30933439 | |