siRNA Details
| Gene Symbol: Nipsnap3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028529.1 | NP_082805.1 | 7 | 1047 | ||
| GenBank mRNA | AV260630.1 | - | 2 | 283 | ||
| BY465731.1 | - | 4 | 432 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nipsnap3a| Gene Symbol: Nipsnap3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028529.1 | NP_082805.1 | 7 | 1047 | ||
| GenBank mRNA | AV260630.1 | - | 2 | 283 | ||
| BY465731.1 | - | 4 | 432 | |||