siRNA Details
| Gene Symbol: Ccsap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028536.1 | NP_082812.1 | 4 | 2645 | ||
| GenBank mRNA | BC024705.1 | - | 3 | 2302 | 3668303 | |
| BC036300.1 | - | 4 | 2645 | 4501054 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccsap| Gene Symbol: Ccsap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028536.1 | NP_082812.1 | 4 | 2645 | ||
| GenBank mRNA | BC024705.1 | - | 3 | 2302 | 3668303 | |
| BC036300.1 | - | 4 | 2645 | 4501054 | ||