siRNA Details
Gene Symbol: Ccsap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028536.1 | NP_082812.1 | 4 | 2599 | ||
GenBank mRNA | BC024705.1 | - | 3 | 2256 | 3668303 | |
BC036300.1 | - | 4 | 2599 | 4501054 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccsap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028536.1 | NP_082812.1 | 4 | 2599 | ||
GenBank mRNA | BC024705.1 | - | 3 | 2256 | 3668303 | |
BC036300.1 | - | 4 | 2599 | 4501054 |