siRNA Details
| Gene Symbol: Tex35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006825.1 | - | 3 | 179 | ||
| BC099580.1 | - | 3 | 174 | 30920417 | ||
| BC115790.1 | - | 3 | 176 | 40100579 | ||
| BY080014.1 | - | 2 | 74 | |||
| BY706810.1 | - | 3 | 179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex35| Gene Symbol: Tex35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006825.1 | - | 3 | 179 | ||
| BC099580.1 | - | 3 | 174 | 30920417 | ||
| BC115790.1 | - | 3 | 176 | 40100579 | ||
| BY080014.1 | - | 2 | 74 | |||
| BY706810.1 | - | 3 | 179 | |||