siRNA Details
Gene Symbol: 1700066B19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033168.2 | NP_001028340.1 | 2 | 1191 | ||
GenBank mRNA | AK006900.1 | - | 1417 | |||
AK162297.1 | - | 2 | 1192 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700066B19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033168.2 | NP_001028340.1 | 2 | 1191 | ||
GenBank mRNA | AK006900.1 | - | 1417 | |||
AK162297.1 | - | 2 | 1192 |