siRNA Details
| Gene Symbol: 1700066M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028546.1 | NP_082822.1 | 2 | 2784 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700066M21Rik| Gene Symbol: 1700066M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028546.1 | NP_082822.1 | 2 | 2784 | ||